missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ inoLab™ Multiparameter Digital Benchtop Meters
Obtain uncompromised measuring accuracy of digital measurements of pH, ORP, dissolved oxygen (optical), BOD, conductivity and turbidity in the lab using the WTW™ inoLab™ Multiparameter Digital Benchtop Meters.
1290.00€ - 2280.00€
Specifications
Outputs | Mini USB |
---|---|
Accuracy (Conductivity) | ±0.05% of reading |
Accuracy (DO) | ±0.05% of reading |
Accuracy (mV) | ±0.3 or 0.05% of reading |
Accuracy (pH) | ±0.005 |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12623800
|
WTW™
1FD35C |
Set including IDS sensors, meter with universal power supply, stand and operation manual, digital IDS pH electrode SenTix™ 940, buffer 4, 7 and 10.01, 3mol/L KCl, digital IDS conductivity cell TetraCon™ 925, 0.01mol/L KCl, conductivity standard, software and USB cable |
2140.00€
Each |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
13593940
|
WTW™
1FD350 |
Meter with universal power supply, stand and operation manual, software and USB cable |
1290.00€
Each |
In Stock
Log in to see stock. |
|||||
12636637
|
WTW™
1FD354 |
Meter with universal power supply, stand and operation manual, optical IDS D.O. sensor FDO™ 925, software and USB cable |
2280.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13503950
|
WTW™
1FD351 |
Set including IDS sensor, meter with universal power supply, stand and operation manual, digital IDS pH electrode SenTix™ 940, buffer 4, 7 and 10.01, 3mol/L KCl, software and USB cable |
1570.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13513950
|
WTW™
1FD352 |
Set including IDS sensor, meter with universal power supply, stand and operation manual, digital IDS pH electrode SenTix™ 980, buffer 4, 7 and 10.01, 3mol/L KCl, software and USB cable |
1600.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13523950
|
WTW™
1FD353 |
Set including IDS sensor, meter with universal power supply, stand and operation manual, digital IDS conductivity cell TetraCon™ 925, 0.01mol/L KCl, conductivity standard, software and USB cable |
1855.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Highly suitable for digital measurements of pH, ORP, dissolved oxygen (optical), BOD, conductivity and turbidity in the lab
- IDS technology allows optimized measurements and efficient documentation in the simplest manner
- Offer the correct solution for pH, ORP, dissolved oxygen and conductivity measurements in the lab
- Uncompromised measuring accuracy
- Wireless ready, digital sensor recognition, smart sensor evaluation
Specifications
Mini USB | |
±0.05% of reading | |
±0.005 | |
1 to 5 | |
500 Manual/5000 automatic with time/date stamp | |
0 μS to 1000 mS | |
-2 to 20.001 | |
0 to 1999 mg/L | |
inoLab™ Multi 9310 IDS | |
Digital benchtop multiparameter | |
3 Significant digits | |
0.1mV | |
3 Significant digits | |
±0.1 | |
0.1 | |
24 W x 19 Hcm | |
19 cm | |
8 kg | |
50/60Hz |
±0.05% of reading | |
±0.3 or 0.05% of reading | |
1 | |
cETLus, CE | |
LCD | |
±1200 mV | |
0 to 70 ppt | |
0.1/0.01/0.001 | |
4 x 1.5V AA type batteries | |
0 to 90mg/L | |
3 Significant digits | |
3 Significant digits | |
3 Significant digits | |
0°C to 50°C | |
3 Years | |
24 cm | |
8 cm | |
100/240V |