Learn More
WTW™ inoLab™ Multiparameter Digital Benchtop Meters
GREENER_CHOICE
Obtain uncompromised measuring accuracy of digital measurements of pH, ORP, dissolved oxygen (optical), BOD, conductivity and turbidity in the lab using the WTW™ inoLab™ Multiparameter Digital Benchtop Meters.
Brand:  WTW™ 1FD354
2344.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12636637
  • 2405.00€ / Each
Explore more special offers
Description
Description
  • Highly suitable for digital measurements of pH, ORP, dissolved oxygen (optical), BOD, conductivity and turbidity in the lab
  • IDS technology allows optimized measurements and efficient documentation in the simplest manner
  • Offer the correct solution for pH, ORP, dissolved oxygen and conductivity measurements in the lab
  • Uncompromised measuring accuracy
  • Wireless ready, digital sensor recognition, smart sensor evaluation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mini USB
±0.05% of reading
±0.005
1 to 5
500 Manual/5000 automatic with time/date stamp
0 μS to 1000 mS
-2 to 20.001
0 to 1999 mg/L
Meter with universal power supply, stand and operation manual, optical IDS D.O. sensor FDO™ 925, software and USB cable
4 x 1.5V AA type batteries
0 to 90mg/L
3 Significant digits
3 Significant digits
3 Significant digits
0°C to 50°C
3 Years
24 cm
8 cm
100/240V
±0.05% of reading
±0.3 or 0.05% of reading
1
cETLus, CE
LCD
±1200 mV
0 to 70 ppt
0.1/0.01/0.001
inoLab™ Multi 9310 IDS
Digital benchtop multiparameter
3 Significant digits
0.1mV
3 Significant digits
±0.1
0.1
24 W x 19 Hcm
19 cm
8 kg
50/60Hz