Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Brand:  Smeg™ LM40
 View more versions of this product
Product Code. 12351763
  • 1950.00€ / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Description
  • For use with Smeg glassware washers
  • Available with one, two and mixed spraying/injection levels
  • Clean and sanitize a wide variety of labware
Specifications
Specifications
Stainless Steel
40-position flask washing trolley in stainless steel with forty U4140 spigots
Laboratory glassware washers
1 Each
40 items
For washing and drying narrow-necked Glassware up to 490mm height, flasks, round-bottom flasks and g
40 U4140 spigots, without drying system connection
Product Title
Smeg™ Injection Trolley for Laboratory Glassware Washers > For use with: SMEG 60cm washer