Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Brand:  Smeg™ LM24
 View more versions of this product
Product Code. 11995664
  • 1625.00€ / Each
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Smeg glassware washers
  • Available with one, two and mixed spraying/injection levels
  • Clean and sanitize a wide variety of labware
Specifications
Specifications
Stainless Steel
24 items
Injection trolley with one injection level
Glassware washers
1 Each
Injection Trolley
Capacity of 24 items; can wash both large items and small flasks or bottles. Maximum height of glassware is 500mm
Washing narrow-necked glassware of different sizes
Fittings for ULB 40 adaptor nozzles for pipettes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Smeg™ Injection Trolley for Laboratory Glassware Washers > Model LM24; Holds: 24 items; For washing narrow-necked glassware up to 490mmH