Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Brand: Smeg™ LM20DS
This item is not returnable.
View return policy
Description
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifications
Stainless Steel | |
20 items | |
Injection trolley with one spraying/injection level | |
Glassware washers | |
1 Each |
Injection Trolley | |
20-position universal flask washing trolley in stainless steel with 20 spigots and drying system connection. Half the trolley space is free for mixed glassware suitable for a ½ basket accessory. | |
Suitable for washing and drying narrownecked glassware up to 19.3 inches / 490 mm in height. | |
20 spigots, drying system connection |
Product Content Correction
Product Title
Smeg™ Injection Trolley for Laboratory Glassware Washers > Model LM20DS; Holds: 20 items; For washing and drying narrow-necked glassware (flasks, round-bottom flasks and graduated cylinders of various sizes)up to 490mmH