Learn More
Thermo Scientific™ Impeller Sleeve and Connector for Drive Shaft DS300
The Impeller sleeve for open-top mixing supports mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPCs) for Thermo Scientific™ HyPerforma™ Single-Use Mixer, with impeller-based mixing in open or closed designs.
Brand:  Thermo Scientific™ SH30749.13
 View more versions of this product
Product Code. 30040285
  • 2345.00€ / Pack of 7
Explore more special offers
Description
Description
  • For use with the open top BPC tank liner and re-usable hub.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 L
108 cm (43 in.) Drive Shaft
Impeller Sleeve
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Impeller Sleeve and Connector for Drive Shaft DS300 > Capacity: 200 L, For Use With: 108 cm (43 in.) Drive Shaft