Learn More
Brady™ Universal Multi-Pole Breaker Lockout
Compatible with most two or three pole breakers with a tie-bar such as GE ITE, Sylvania, Challenger, Bryant, Westinghouse and General Switch Company.
Brand: Brady™ 066320
This item is not returnable.
View return policy
Description
- Use thumbscrews to clamp lockout securely over tie-bar, then insert lock to prevent clamp from being loosened
Specifications
Universal Lock for Multi-pole Circuit Breaker | |
Glass-filled Nylon | |
Universal Multi-Pole Breaker Lockout | |
31.8 mm | |
62.2 mm |
Red/Black | |
UL 94 HB | |
62.20 x 31.80 mm | |
6 Pk. |
Product Content Correction
Product Title
Brady™ Universal Multi-Pole Breaker Lockout > Width: 62.2mm