missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva IMAC Sepharose™ 6 Fast Flow Affinity Media
Used for purifying proteins and peptides that have an affinity for metal ions. Cytiva IMAC Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix.
335.00€ - 1114.00€
Specifications
For Use With (Application) | For Purifying Proteins and Peptides That Have an Affinity for Metal Ions |
---|---|
Product Type | IMAC Sepharose 6 Fast Flow Media |
Product Code | Brand | Particle Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Quantity | Price | Quantity & Availability | ||||
10372810
|
Cytiva
17-0921-08 |
45 to 165 μm | 100 mL |
1114.00€
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762842
|
Cytiva
17-0921-07 |
90 μm | 25 mL |
335.00€
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Uncharged IMAC resin allows possibility to charge with metal ion of choice for optimized selectivity
- Used for ions such as histidine-tagged proteins but also untagged recombinant or native proteins
- High protein binding capacity
- Coupling chemistry and novel ligand designed to give minimal ligand leakage
- Available HiTrap™ and HiPrep™ pre-packed formats for easy scale-up and process development
Specifications
For Purifying Proteins and Peptides That Have an Affinity for Metal Ions | |
IMAC Sepharose 6 Fast Flow Media |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- IMAC sepharose 6 fast flow
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva IMAC Sepharose™ 6 Fast Flow Affinity Media