Learn More
IL4R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536394
Description
This antibody detects endogenous protein at a molecular weight of 88 kDa. Purity is >95% by SDS-PAGE.
IL4R Polyclonal antibody specifically detects IL4R in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Specifications
IL4R | |
Polyclonal | |
Unconjugated | |
IL4R | |
582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16190, 25084, 3566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P16382, P24394, Q63257 | |
IL4R, Il4ra | |
Synthetic peptide corresponding to amino acids 460-510 of human IL-4R alpha | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.