missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IKK gamma Inhibitor Peptide Set
Applications: In vitro assay, In vivo assay, Functional (Inhibition), Block/Neutralize
579.00€ - 1105.00€
Specifications
Host Species | Human, Mouse, Rat |
---|---|
Components | 1.IKK-gamma NEMO Binding Domain (NBD) Inhibitor Peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKTALDWSWLQTE (IKK-gamma/NEMO binding sequence is underlined). Molecular weight: 3692 2.Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 |
For Use With (Application) | Functional (Inhibition), In vitro assay, In vivo assay |
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Molecular Weight (g/mol) | 3692 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18114365
|
Novus Biologicals™
NBP2-26504-5MG |
5 mg |
1105.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18780644
|
Novus Biologicals™
NBP2-26504 |
579.00€
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Specifications
Human, Mouse, Rat | |
Functional (Inhibition), In vitro assay, In vivo assay | |
3692 |
1.IKK-gamma NEMO Binding Domain (NBD) Inhibitor Peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKTALDWSWLQTE (IKK-gamma/NEMO binding sequence is underlined). Molecular weight: 3692 2.Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 |
|
Store at -20°C. Avoid freeze-thaw cycles. | |
IKK gamma Inhibitor |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ IKK gamma Inhibitor Peptide Set