missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF9 Antibody (993107) [Janelia Fluor™ 646], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9140J
This item is not returnable.
View return policy
Description
IGSF9 Monoclonal antibody specifically detects IGSF9 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
IGSF9 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57549 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
993107 | |
Flow Cytometry, CyTOF-ready | |
FP18798, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGSF9, Arg21-Leu730, Accession # Q05BQ1 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IGSF9 Antibody (993107) [Janelia Fluor™ 646], Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
Spot an opportunity for improvement?Share a Content Correction