missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Igepal Ca - 630 Non-Ionic Detergent MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Spezifikation
Spezifikation
Density | 1.06 g/mL |
Quantity | 50 mL |
Chemical Name or Material | Igepal Ca - 630, Non-Ionic Detergent |
Produktvorschläge
Customers who viewed this item also viewed
Sicherheit und Handhabung


Produktidentifikator
- Igepal CA-630
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Igepal Ca - 630 Non-Ionic Detergent MP Biomedicals >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur