missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IB-MECA, Tocris Bioscience™
Click to view available options
Quantity:
5 mg
25 mg
Description
Potent and selective A3 adenosine receptor agonist (Ki values are 1.1, 54 and 56 nM for A3, A1 and A2A receptors respectively). Cardioprotective, reduces infarct size upon reperfusion in rats.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Quantity | 5 mg |
Formula Weight | 510.29 |
Percent Purity | >97% |
Chemical Name or Material | IB-MECA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IB-MECA, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction