missing translation for 'onlineSavingsMsg'
Learn More
[Hydroxy(tosyloxy)iodo]benzene, 97%, Thermo Scientific Chemicals Product Code.: 10035293

[Hydroxy(tosyloxy)iodo]benzene, 97%, Thermo Scientific Chemicals

Product Code. 10035293
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10035293

Brand: Thermo Scientific Acros 389610250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27126-76-7
Molecular Formula C13H13IO4S
Molecular Weight (g/mol) 392.21
MDL Number MFCD00011547
InChI Key LRIUKPUCKCECPT-UHFFFAOYSA-N
Synonym hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
PubChem CID 325434
IUPAC Name [hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 131.0°C to 137.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 5369
Formula Weight 392.21
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material [Hydroxy(tosyloxy)iodo]benzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450973 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number389610250,389610050
Lot NumberA0462187 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number389610250,389610050
Lot NumberA0462374 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number389610250,389610050
Lot NumberA0450972 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number389610250,389610050
Lot NumberA0450973 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number389610250,389610050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
[Hydroxy(tosyloxy)iodo]benzene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.