missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
Description
Hydrogen Peroxide, 30% (Certified ACS)

Specifications
Specifications
Color | Colorless |
Boiling Point | 108°C |
Melting Point | -33°C |
Assay Percent Range | 29.0 to 32.0 % |
Density | 1.110g/cm³ |
Formula Weight | 34 |
Percent Purity | 29.0 to 32.0% |
Grade | ACS |
Identification | Pass test |
Packaging | Poly Bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction