missing translation for 'onlineSavingsMsg'
Learn More
Hydrofluoric acid, Honeywell™ Product Code.: 16655853

Hydrofluoric acid, Honeywell™

Product Code. 16655853 Shop All Honeywell Research Chemicals Products
5 L, Plastic Bottle
Click to view available options
Quantity:
5 L
Packaging:
Plastic Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16655853

Brand: Honeywell Research Chemicals 40213H5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Semiconductor grade (Honeywell 17736), 49.5-50.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7664-39-3, 7732-18-5
Boiling Point 105°C (1.013 hPa)
CAS Min % ≥40.00%
CAS Max % <60.00%
Packaging Plastic Bottle
Quantity 5 L
Molecular Formula FH
Physical Form Liquid
MDL Number MFCD00011346
Beilstein 1098214
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES F
IUPAC Name hydrogen fluoride
Molecular Weight (g/mol) 20.01
PubChem CID 14917
ChEBI CHEBI:29228
Grade Semiconductor grade acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrofluoric acid, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.