missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (R338), DyLight 405 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89761V
This item is not returnable.
View return policy
Description
Human Respiratory Syncytial Virus A2 Fusion Protein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Human Respiratory Syncytial Virus A2 Fusion Protein | |
Monoclonal | |
DyLight 405 | |
HRSVgp08, RSV, RSV fusion protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (RSV) Fusion glycoprotein/ RSV-F (AAB59858.1; Met1-Thr529) and was produced using recombinant antibody technology. | |
0.2 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
R338 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (R338), DyLight 405 , Novus Biologicals™ > 0.2 mL; DyLight 405
Spot an opportunity for improvement?Share a Content Correction