missing translation for 'onlineSavingsMsg'
Learn More

Human Perforin Antibody, R&D Systems™

Rabbit Monoclonal Antibody

222.00€ - 564.00€

Specifications

Antigen Perforin
Clone 2771C
Dilution Immunohistochemistry 5-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18734127
View Documents
R&D Systems
MAB11298-SP
25 μg
222.00€
25µg
Please to purchase this item. Need a web account? Register with us today!
18730177
View Documents
R&D Systems
MAB11298-100
100 μg
564.00€
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Immunohistochemistry
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Human Perforin Antibody, R&D Systems™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.