Learn More
R&D Systems™ Human PDGF-CC Quantikine ELISA Kit
a 4.5 hour solid phase ELISA designed to measure human PDGF-CC in cell culture supernates, serum, and platelet-poor plasma.
Brand: R&D Systems™ DCC00
This item is not returnable.
View return policy
Description
The Human PDGF-CC Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PDGF-CC in cell culture supernates (50 ul), serum (50 ul), platelet-poor edta plasma (50 ul), platelet-poor heparin plasma (50 ul).
Specifications
62.50 - 4000pg/mL | |
HRP | |
Natural and Recombinant Human PDGF-CC | |
ELISA | |
Cell Culture Supernates (50uL),Serum (50uL),Platelet-poor EDTA Plasma (50uL),Platelet-poor Heparin Plasma (50uL) |
24.7pg/mL | |
Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Heparin), Serum | |
1 x 96-well Plate | |
56034 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
R&D Systems™ Human PDGF-CC Quantikine ELISA Kit > One 96-well plate