Learn More
R&D Systems™ Human PDGF-CC Quantikine ELISA Kit
a 4.5 hour solid phase ELISA designed to measure human PDGF-CC in cell culture supernates, serum, and platelet-poor plasma.
Brand:  R&D Systems™ DCC00
Product Code. 18771974
  • 835.00€ / 1 set
Explore more special offers
Description
Description
The Human PDGF-CC Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PDGF-CC in cell culture supernates (50 ul), serum (50 ul), platelet-poor edta plasma (50 ul), platelet-poor heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
62.50 - 4000pg/mL
HRP
Natural and Recombinant Human PDGF-CC
ELISA
Cell Culture Supernates (50uL),Serum (50uL),Platelet-poor EDTA Plasma (50uL),Platelet-poor Heparin Plasma (50uL)
24.7pg/mL
Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Heparin), Serum
1 x 96-well Plate
56034
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Human PDGF-CC Quantikine ELISA Kit > One 96-well plate