missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human M-CSF Quantikine ELISA Kit

Human M-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

752.00€ - 4290.00€

Specifications

Assay Range 78.10 to 5000 pg/mL
Assay Sensitivity 47.3 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
View More Specs

Products 2
Product Code Brand Quantity Sufficient For Price Quantity & Availability  
Product Code Brand Quantity Sufficient For Price Quantity & Availability  
18712374
View Documents
R&D Systems™
DMC00B
1 Plate 1 x 96-well Plate
752.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18732104
View Documents
R&D Systems™
SMC00B
6 Plates 6 x 96-well Plates
4290.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Human M-CSF Quantikine ELISA Kit quantifies human M-CSF.
Specifications

Specifications

78.10 to 5000 pg/mL
HRP
Solid Phase Sandwich ELISA
Natural and Recombinant Human M-CSF
ELISA
Cell Culture Supernates (100 μL), Serum (100 μL), EDTA Plasma (100 μL), Heparin Plasma (100 μL), Saliva (100 μL), Urine (100 μL)
colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
47.3 pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Human
1435
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Human M-CSF Quantikine ELISA Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.