Learn More
R&D Systems™ Human LRRC3B (NP_443185) VersaClone cDNA
Leucine Rich Repeat Containing 3B
Brand:  R&D Systems™ RDC1769
Product Code. 18719218
  • 322.00€ / 10µg
Explore more special offers
Description
Description
The Human LRRC3B (NP_443185) VersaClone cDNA clone contains the complete ORF for LRRC3B/LRP15, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LRRC3B/LRP15 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cDNA
NP_443185
cDNA is provided in 10mM Tris-Cl, pH 8.5
116135 (Human), 218763 (Mouse)
Human
hLRRC3B
10 μg at 0.2 μg/μL
792bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Human LRRC3B (NP_443185) VersaClone cDNA > Quantity: 10μg