Learn More
Gibco™ Human GM-CSF Recombinant Protein, PeproTech®
Recombinant Protein
83.75€ - 3360.00€
Specifications
Accession Number | P04141 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 1437 |
Molecular Weight (g/mol) | 14.6 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17850053
|
Gibco™
300-03-100UG |
100 μg |
674.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17851803
|
Gibco™
300-03-1MG |
1 mg |
3360.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831823
|
Gibco™
300-03-20UG |
20 μg |
203.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17871803
|
Gibco™
300-03-250UG |
250 μg |
1180.00€
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830533
|
Gibco™
300-03-500UG |
500 μg |
2010.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861303
|
Gibco™
300-03-50UG |
50 μg |
402.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880033
|
Gibco™
300-03-5UG |
5 μg |
83.75€
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant Human GM-CSF is a 14.6 kDa globular protein consisting of 128 amino acids, containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.Specifications
P04141 | |
protein with no preservative | |
14.6 kDa | |
E. coli | |
<1 EU/ μg | |
GM-CSF | |
The ED50 as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg | |
Recombinant | |
-20°C | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
1437 | |
Human GM-CSF | |
RUO | |
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim | |
CSF2 | |
Unconjugated | |
MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE | |
E. coli | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.