missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
220.00€ - 5630.00€
Specifications
Accession Number | P21781 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2252 |
Molecular Weight (g/mol) | 18.9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17813633
|
Gibco™
AF-100-19-100UG |
100 μg |
1170.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17823803
|
Gibco™
AF-100-19-10UG |
10 μg |
220.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17863073
|
Gibco™
AF-100-19-1MG |
1 mg |
5630.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P21781 | |
protein with no preservative | |
18.9 | |
E. coli | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2252 | |
Human FGF-7 (KGF), Animal-Free | |
-20°C | |
<0.1 EU/μg | |
FGF7 | |
Unconjugated | |
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |