missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ AF-100-19-1MG
This item is not returnable.
View return policy
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P21781 | |
protein with no preservative | |
18.9 | |
1 mg | |
-20°C | |
<0.1 EU/μg | |
FGF7 | |
Unconjugated | |
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
2252 | |
Human FGF-7 (KGF), Animal-Free | |
E. coli | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Human | |
Recombinant | |
Lyophilized |