Learn More
Novus Biologicals™ Horse Serum Albumin Native Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP2-50615
This item is not returnable.
View return policy
Description
Native Horse Serum Albumin. Source: Serum The Horse Serum Albumin Native Protein has been validated for the following applications: Western Blot, ELISA.
Specifications
P35747 | |
0.01M Sodium Phosphate, 0.14M Sodium Chloride, pH 7.4 | |
10 mg | |
ALB Equ c 3, Serum Albumin | |
Unconjugated | |
Multi-step |
ELISA, Western Blot | |
100034206 | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Horse Serum Albumin | |
Lyophilized |
Product Content Correction
Product Title
Novus Biologicals™ Horse Serum Albumin Native Protein > 10mg