Learn More
Novus Biologicals™ Horse Serum Albumin Native Protein
Highly purified. Generating reliable and reproducible results.
Brand:  Novus Biologicals™ NBP2-50615
Product Code. 18188267
  • 314.00€ / 10mg
Explore more special offers
Description
Description
Native Horse Serum Albumin. Source: Serum The Horse Serum Albumin Native Protein has been validated for the following applications: Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P35747
0.01M Sodium Phosphate, 0.14M Sodium Chloride, pH 7.4
10 mg
ALB Equ c 3, Serum Albumin
Unconjugated
Multi-step
ELISA, Western Blot
100034206
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Horse Serum Albumin
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Horse Serum Albumin Native Protein > 10mg