missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Buffer Chamber

For use with TE62 Standard Transfer Tank with Cooling Chamber

Brand:  Hoefer™ TE67

 View more versions of this product

Product Code. 11584444

  • 1595.00€ / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With Heat Exchanger
Specifications

Specifications

TE 62 or the discontinued TE 62X Transfer Tank
Buffer Chamber
Fittings, crochet hook and holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions