missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ High-Purity Cyclohexyl Maltoside Detergents

Cyclohexyl maltoside detergents are a class of specialized non-ionic detergents designed for the solubilization, stabilization, and manipulation of biomolecules, particularly membrane proteins.

216.00€ - 1030.00€

Specifications

Color White
Product Type Detergent
Solubility ≥20% (in water at 20°C)
Content And Storage Store below -20°C.
Form Lyophilized
View More Specs

Products 6
Product Code Brand Quantity Reagent Type Price Quantity & Availability  
Product Code Brand Quantity Reagent Type Price Quantity & Availability  
30245344
View Documents Promotion Details
Thermo Scientific™
A65517
1 g Cymal-5
216.00€
1g
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
30245087
View Documents
Thermo Scientific™
A65518
5 g Cymal-5
742.00€
5g
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
30245136
View Documents
Thermo Scientific™
A65520
1 g Cymal-6
240.00€
1g
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
30245146
View Documents
Thermo Scientific™
A65521
5 g Cymal-6
910.00€
5g
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
30245343
View Documents
Thermo Scientific™
A65522
1 g Cymal-7
288.00€
1g
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
30245216
View Documents
Thermo Scientific™
A65523
5 g Cymal-7
1030.00€
5g
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
Description
Show More
Specifications

Specifications

White
≥20% (in water at 20°C)
Lyophilized
Detergent
Store below -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ High-Purity Cyclohexyl Maltoside Detergents

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.