missing translation for 'onlineSavingsMsg'
Learn More

Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB110-68095DL594

Product Code. 18502239

  • 574.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Hepatic Sinusoidal Endothelial Cells Monoclonal antibody specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hepatic Sinusoidal Endothelial Cells
Monoclonal
DyLight 594
Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells
Rat Hepatic Sinusoidal Endothelial Cells
0.1 mL
Endothelial Cell Markers
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
SE-1
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Store at 4°C in the dark.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), DyLight 594, Novus Biologicals™ > 0.1 mL; DyLight 594

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.