Learn More
Eppendorf™ Heat Sealer S200
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
Brand:  Eppendorf™ 5392000005
 View more versions of this product
Product Code. 15343499
  • 4492.90€ / Each
Explore more special offers
Description
Description
  • Eliminates evaporation in PCR
  • Adjustable sealing temperatures for more flexibility
  • Seals 96- and 384-well plates
  • Sealing time selectable from 1 to 9 seconds in 0.5 second increments
  • Integrated thermostat prevents overheating
  • Powerful handle for ergonomic operation
  • PCR plate adapter included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/60 Hz
220 × 321 × 425 mm
96- and 384-well plates
125°C to 200°C in 1°C increments
100/230 V
100/230 V, 50/60 Hz
EU Plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Heat Sealer S200 > EU Plug