Learn More
Eppendorf™ Heat Sealer S100
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
3132.48€
Specifications
Hertz 50/60 Hz
Voltage 100/230 V
Dimensions (L x W x H) 220 × 250 × 230 mm
Electrical Requirements 100/230 V, 50/60 Hz
For Use With (Equipment) 96- and 384-well plates
View More Specs
Products 1
Product Code Brand Plug Type Price Quantity & Availability  
Product Code Brand Plug Type Price Quantity & Availability  
15530566
View Documents
Eppendorf™
5391000001
EU Plug
3132.48€
Each
Description
Description
  • Hermetic sealing of multiwell plates
  • Fixed sealing temperature for reproducible results
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Compact design, portable device
  • Integrated thermostats prevents overheating
  • Improved mechanics for even easier sealing
  • PCR plate adapter included
Specifications
Specifications
50/60 Hz
220 × 250 × 230 mm
96- and 384-well plates
100/230 V
100/230 V, 50/60 Hz
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ Heat Sealer S100