Learn More
asecos™ Hazardous Substances Expandable Rack
The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.
549.00€ - 1115.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17268450
|
asecos™
27838 |
20 L |
549.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17866526
|
asecos™
27842 |
25 L |
612.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17836526
|
asecos™
27840 |
30 L |
563.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17826556
|
asecos™
27846 |
32 L |
643.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17854625
|
asecos™
27844 |
40 L |
679.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17816436
|
asecos™
27850 |
41 L |
781.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17836356
|
asecos™
27848 |
45 L |
715.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17874625
|
asecos™
27852 |
57 L |
891.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17896556
|
asecos™
27866 |
205 L |
1115.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Flat-pack delivery with assembly instructions
- Galvanised grids as storage levels and 1 bottom collecting sump made of galvanised steel.
Specifications
Steel | |
Drum Rack |