missing translation for 'onlineSavingsMsg'
Learn More

Schmidlin™ GT Plus 6000 Ultra Zero Air Generator

This generator produces a continuous supply of dry, ultra zero grade air.

Brand:  Schmidlin™ DB-NGT30000-EU

Encompass_Preferred

 View more versions of this product

Product Code. 17768909

  • 14048.03€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Purity: Free from THC, CO CO2, O3, NOx and SOx
  • Pressure: 4.5 to 10 barg (65 to 145 psig)
  • Proprietary platinum catalyst technology
  • Flow capacity to match your specific instrument demands
  • Meets and exceeds requirements for the most demanding analyzers
Specifications

Specifications

31 cm
47 cm
3.5 to 9 bar
63 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
565 W
30,000 mL/min.
35°C
25 in.
Inlet: 1/4 in., Outlet: 1/8 in.
<0.1 ppm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.