Learn More
Tocris Bioscience™ GSK 626616
Potent and selective DYRK inhibitor; orally bioavailable.
Brand: Tocris Bioscience™ 6638/10
224.62 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
GSK 626616 is a potent and selective DYRK inhibitor (IC50 = 0.7 nM for DYRK3, similar potency for DYRK1 and 2). Exhibits ∽20-fold selectivity over casein kinase 2 and negligible activity against a panel of 451 kinases screened. Enhances number of CFU-E stimulated by Epo from human marrow. Increases hemoglobin levels in anemic mice. Orally bioavailable.Specifications
(5Z)-2-[(2,6-Dichlorophenyl)amino]-5-(6-quinoxalinylmethylene)-4(5H)-thiazolone | |
10 mg | |
401.27 |
1025821-33-3 | |
C18H10Cl2N4OS | |
98% |