Learn More
Greiner Bio-One VACUETTE™ CAT Serum Clot Activator Blood Collection Tubes
Click to view available options
Capacity (Metric):
2 mL
4 mL
6 mL
9 mL
Unit Size:
Pack of 1200
Pack of 50
Description
- VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
- Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
- VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
- Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
- Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
- Sterile and latex-free.
Specifications
Specifications
| Color | Transparent |
| Product Type | Blood Collection Tube |
| Additive | Clot Activator |
| Material | PET |
| Sterility | Sterile |
| Closure Type | Non-ridged Pull Cap |
| Capacity (Metric) | 6 mL |
| Diameter (Metric) | 13 mm |
| Closure Color | Red with Black Ring |
| Tube Size | 13 x 100 mm |
| Show More |