Learn More
Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry
38.00€ - 76.44€
Specifications
Additive | Lithium Heparin |
---|---|
Closure Type | Safety Pull Cap |
Closure Color | Green with Yellow Ring |
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
12046649
|
Greiner Bio-One
455083 |
8 mL | 16 x 100 mm | N/A | |||||
15442520
|
Greiner Bio-One
454008 |
4 mL | 13 x 75 mm |
38.00€
Pack of 50 |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
12036649
|
Greiner Bio-One
456087 |
5 mL | 13 x 100 mm | N/A | |||||
Description
- Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
- Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Nonscrew Safety Pull Caps can be removed with a gentle pull
Specifications
Lithium Heparin | |
Safety Pull Cap | |
Green with Yellow Ring |