missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Highly potent and selective κ-opioid agonist (IC50 = 0.04 nM) that may be selective for the putative κ2 receptor. Anti-nociceptive and neuroprotective in vivo.

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 530.4 |
Percent Purity | >99% |
Chemical Name or Material | GR 89696 fumarate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GR 89696 fumarate, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction