Learn More
Gosselin™ Polystrene Petri Dish Product Code.: 11718593
Only undefined left
Add to basket
Gosselin™ Polystrene Petri Dish
Product Code. 11718593
140 mm, 20.6 mm, 0, Pack of 176
Click to view available options
Diameter (Metric):
55 mm
140 mm
Height (Metric):
14.2 mm
20.6 mm
Vent:
0
3
6
Unit Size:
Pack of 1620
Pack of 176
Product Code. 11718593
Brand: Gosselin BP14002
Only undefined left
Add to basket
Only undefined left
Add to basket
Crystal polystyrene dishes, with optimal flatness.
  • Appropriate for aseptic production
  • Sterilized by irradiation
  • Flat bottom
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC N°1935/2004, ISO 11137
Color Natural
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
Well Count 1
For Use With (Application) For use in clinical, environment, industries, microbiology
Quantity 176 Carton
Vent 0
Height (Metric) 20.6 mm
Diameter (Metric) 140 mm
Product Title
Gosselin™ Polystrene Petri Dish >