missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Watson-Marlow Tubing GORE™ STA-PURE PFL Tubing
GORE™ STA-PURE PFL Tubing, formerly known as Chem-sure tubing, handles nearly all aggressive chemicals, including organic solvents such as methyl ethyl ketone, toluene, and acetone.
Brand: Watson-Marlow Tubing 966.0080.016
This item is not returnable.
View return policy
Description
- STA-PURE PFL handles nearly all aggressive chemicals, including organic solvents such as methyl ethyl ketone, toluene, and acetone.
- STA-PURE PFL is a high-performance composite of expanded polytetrafluoroethylene (ePTFE) and a platinum-cured perfluoroelastomer, giving extraordinary chemical resistance, extremely long life and very high burst pressures.
- It is free from plasticisers, acid acceptors and other processing aids, making it one of the purest tubings available.
- Leachability tests using ethyl acetate yielded over 100 times less total extractables than other fluoroelastomer tubing materials.
- In addition, STA-PURE PFL has Class VI tests for pharmaceutical applications.
Specifications
PFL Tubing | |
0.3125 in. | |
ePTFE, Platinum-cured FFKM | |
305 mm | |
8.0 mm | |
85 (Shore A, ± 10) | |
80°C |
Autoclavable | |
8.0 mm | |
12 in. | |
0.3125 in. | |
Off-white | |
176°F |