Learn More
GPE Scientific™ Glass Potter Tissue Grinders
117.00€ - 142.00€
Specifications
Clearance (Metric) | 0.152 - 0.254mm |
---|---|
Autoclavable | Autoclavable |
Material | Glass |
Product Type | Tissue Grinder |
Pestle Material | Glass |
Specifications
0.152 - 0.254mm | |
Glass | |
Glass |
Autoclavable | |
Tissue Grinder |