Learn More
GPE Scientific™ Glass Potter Tissue Grinders
117.00€ - 142.00€
Specifications
Clearance (Metric) 0.152 - 0.254mm
Autoclavable Autoclavable
Material Glass
Product Type Tissue Grinder
Pestle Material Glass
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11726923
View Documents Promotion Details
GPE Scientific™
20716F
5 mL N/A
N/A
11721284
View Documents Promotion Details
GPE Scientific™
20718F
15 mL N/A
N/A
11731284
View Documents Promotion Details
GPE Scientific™
20719F
30 mL N/A
N/A
Specifications
0.152 - 0.254mm
Glass
Glass
Autoclavable
Tissue Grinder