Learn More
GPE Scientific™ Glass Potter Tissue Grinders
Brand: GPE Scientific™ 20719F
This item is not returnable.
View return policy
Specifications
0.152 - 0.254mm | |
30 mL | |
Tissue Grinder | |
Glass |
Autoclavable | |
Glass | |
105mm |