Learn More
Rogo Sampaic™ Glass Hellendahl Staining Jar
Holds 8 microscope slides of approx. 76 x 26mm or for 16 slides back-to-back. Rogo Sampaic™ Glass Hellendahl Staining Jars are manufactured from soda lime glass.
Brand:  Rogo Sampaic™ BPL001
Product Code. 11712654
  • 38.48€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Clear glass
  • High wall shape
Specifications
Specifications
Glass
8
60 mm
100 mm
Staining Jar
60 x 60 mm
60 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Rogo Sampaic™ Glass Hellendahl Staining Jar > Slide Count: 8 Slides