Learn More
Sartorius™ Glass Funnels
Use with glass vacuum filter holder assemblies. Sartorius™ Glass Funnels are made of high-quality borosilicate glass. Available in 30, 250 and 400mL capacities.
Brand:  Sartorius™ 6980115
64.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10273491
  • 98.35€ / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Description
  • Available in three sizes for use with various filter sizes and filtration applications
  • Constructed of autoclavable borosilicate glass
  • For use in glass vacuum filter holder assemblies
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 mL
Borosilicate Glass
Filtration with glass vacuum filter holder assemblies
Glass Funnel
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    22
    Hours
    0
    Minutes
    17
    Seconds
    46
Product Content Correction
Product Title
Sartorius™ Glass Funnels > Funnel