Learn More
RETSCH Glass Beads for Mills
For use with Retsch MM 200 Mixer Mills.
Brand:  RETSCH 22.222.0005
 View more versions of this product
Product Code. 10782701
  • 57.95€ / 500g
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Description
  • Ideal for disruption of biological cells
Specifications
Specifications
Beads
1 to 1.5 mm
500 g
Glass
Mixer Mill MM 200
Documents
One moment while we fetch your results.
Product Title
RETSCH Glass Beads for Mills > Diameter: 1 to 1.5mm