Learn More
Genevac™ miVac Centrifugal Concentrators
Choose the ideal concentrator according to workload. Genevac™ miVac Centrifugal Concentrators are modular systems for removing water and solvents from samples. They are available with a range of unique JetRotors™ and vacuum pumps. An optional high-power cold trap is used to condense solvent vapors.
Brand:  Genevac™ DUC-23050-B00
 View more versions of this product
Product Code. 10110113
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Show More
Specifications
Specifications
miVac Duo
Samples in tubes, microplates and vials
Concentrator with small concentration chamber
360W x 424D x 300H mm
230 V 50/60 Hz 650 w
30°C to 80°C
120 Reaction Vessels (0.5 mL), 6 Microtiter Plates
Max G-force: 250
21 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Product Information
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Genevac™ miVac Centrifugal Concentrators > Model: miVac Duo; Capacity: 120 reaction vessels 0.5mL or 6 microtiter plates; Includes concentrator with small chamber