Learn More
Thermo Scientific™ General Gas Chromatography Tubing and Tools
Use Thermo Scientific™ General Gas Chromatography Tubing and Tools to maintain and optimize your GC instrument.
7.18€ - 442.56€
Specifications
For Use With (Application) GC
View More Specs
Products 8
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10169534
View Documents
Thermo Scientific™
60180-770
Straight-Tipped Forceps N/A N/A N/A
10037364
View Documents
Thermo Scientific™
60180-771
Angle-Tipped Forceps N/A N/A N/A
10468014
View Documents
Thermo Scientific™
60180-779
0.35 mm Drill Bit N/A N/A N/A
10047494
View Documents
Thermo Scientific™
60180-780
0.45 mm Drill Bit N/A N/A N/A
10392835
View Documents
Thermo Scientific™
60180-781
0.70 mm Drill Bit N/A N/A N/A
10200545
View Documents
Thermo Scientific™
60180-835
Shortix Capillary Column Cutter
442.56€
Each
In Stock
to see stock.
Add to basket
10548995
View Documents
Thermo Scientific™
60180-836
Shortix Capillary Column Cutter Repair Kit
234.82€
Each
In Stock
to see stock.
Add to basket
10097724
View Documents Promotion Details
Thermo Scientific™
60201-318
Ceramic Column Cutter
7.18€
Each
In Stock
to see stock.
Add to basket
Description
Description
Use Thermo Scientific™ General Gas Chromatography Tubing and Tools to maintain and optimize your GC system and support your GC applications.
Specifications
Specifications
GC
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ General Gas Chromatography Tubing and Tools