Learn More
RETSCH Gasket For Grinding Jar
Brand:  RETSCH 03.111.0289
 View more versions of this product
Product Code. 15310984
  • 18.10€ / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Specifications
For 50mL grinding jar
1 Pack
Gasket
50 mL Grinding Jar
Stainless Steel, PTFE
Product Title
RETSCH Gasket For Grinding Jar > For 50mL Grinding Jar, 1 pack