Learn More
Usbeck Laborgeraete™ Gas Lighters
Brand:  Usbeck Laborgeraete™ 1640
 View more versions of this product
Product Code. 10119270
  • 3.81€ / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Specifications
155 mm
Documents
One moment while we fetch your results.
Product Title
Usbeck Laborgeraete™ Gas Lighters > Length: 155mm