missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GammaBind™ Plus Sepharose™ Medium

Engineered for better binding to rat and mouse IgG subclasses

1043.00€ - 22200.00€

Specifications

Product Type GammaBind Plus Sepharose
Ligand Recombinant GammaBind G type 3 lacking albumin-binding region
Particle Size 90 μm
Content And Storage 2°C to 8°C, 20% Ethanol
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11584945
View Documents
Cytiva
17-0886-01
5 mL
1043.00€
5mL
Please to purchase this item. Need a web account? Register with us today!
11594945
View Documents
Cytiva
17-0886-02
25 mL
3909.00€
25mL
Please to purchase this item. Need a web account? Register with us today!
11504955
View Documents
Cytiva
17-0886-04
500 mL
22200.00€
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
  • More enhanced binding capabilities than GammaBind G at both laboratory and process scale
  • Yields up to 18mg/mL mouse IgG
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse

Specifications

Specifications

GammaBind Plus Sepharose
Recombinant GammaBind G type 3 lacking albumin-binding region
90 μm
2°C to 8°C, 20% Ethanol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • gammabind plus sepharose

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.