missing translation for 'onlineSavingsMsg'
Learn More
FURA-2AM, Tocris Bioscience™ Product Code.: 18054602

FURA-2AM, Tocris Bioscience™

Product Code. 18054602
1 mg
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy

Product Code. 18054602

Brand: Tocris Bioscience 2220/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescent Ca2+ indicator

FURA-2AM is a fluorescent ratiometric Ca2+ indicator. FURA-2AM is selective for Ca2+ over other divalent cations Mg2+, Zn2+, Fe2+ and Mn2+. FURA-2AM binds to free intracellular calcium and is used to determine [Ca2+]i concentration. This product is typically prepared in DMSO. F 127 (Cat. No. 6253) for the solubilization of FURA-2AM is also available.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108964-32-5
Molecular Formula C44H47N3O24
Quantity 1 mg
Synonym fura 2-am, fura-2 pentakis acetoxymethyl ester, bis acetoxymethyl 2,2'-2-5-acetoxymethoxy carbonyl oxazol-2-yl-5-2-2-bis 2-acetoxymethoxy-2-oxoethyl amino-5-methylphenoxy ethoxy benzofuran-6-yl azanediyl diacetate, fura-2 am, fura-2-acetoxymethyl ester, 1-2-5-carboxyoxazol-2-yl-6-aminobenzofuran-5-oxy-2-2′-amino-5′-methylphenoxy-ethane-n,n,n′n′-tetraacetic acid pentaacetoxymethyl ester, 1-6-amino-2-5-carboxy-2-oxazolyl-5-benzofuranyloxy-2-2-amino-5-methylphenoxy ethane-n,n,n',n'-tetraacetic acid, pentaacetoxymethyl ester, fura-2/am, fura-2 acetoxymethyl ester, fura 2-am hplc
InChI Key VPSRLGDRGCKUTK-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOC2=C(C=C3C(=C2)C=C(O3)C4=NC=C(O4)C(=O)OCOC(=O)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C
IUPAC Name acetyloxymethyl 2-[6-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]-1-benzofuran-2-yl]-1,3-oxazole-5-carboxylate
Molecular Weight (g/mol) 1001.857
PubChem CID 3364574
Formula Weight 1001.85
Percent Purity >95%
Chemical Name or Material FURA-2AM
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FURA-2AM, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.