missing translation for 'onlineSavingsMsg'
Learn More

Labconco Freestanding Washer Installation Kit 230V

Washer install kit includes everything for a smooth installation. Kit includes hoses, electrical panel, harnesses and a hose adapter for water. A power cord is required.

Brand:  Labconco 4579801

Product Code. 18095461

  • 240.00€ / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Kit includes:

  • Hoses
  • Electrical panel
  • Harnesses
  • Hose adapter for water

A power cord is required.

Specifications

Specifications

Installation Kit
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco Freestanding Washer Installation Kit 230V > Installation Kit: Glassware Washer; Voltage: 230V Type: Freestanding

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.